siRNA Details
| Gene Symbol: SPRR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006945.4 | NP_008876.3 | 2 | 439 | ||
| XM_017002175.1 | XP_016857664.1 | 2 | 471 | |||
| GenBank mRNA | AK291317.1 | - | 2 | 441 | ||
| M21302.1 | - | 2 | 439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPRR2D| Gene Symbol: SPRR2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006945.4 | NP_008876.3 | 2 | 439 | ||
| XM_017002175.1 | XP_016857664.1 | 2 | 471 | |||
| GenBank mRNA | AK291317.1 | - | 2 | 441 | ||
| M21302.1 | - | 2 | 439 | |||