siRNA Details
| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001565.3 | NP_001556.2 | 2 | 209 | ||
| GenBank mRNA | AA321649.1 | - | 2 | 209 | ||
| BC010954.1 | - | 2 | 174 | 4274617 | ||
| X02530.1 | - | 2 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXCL10| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001565.3 | NP_001556.2 | 2 | 209 | ||
| GenBank mRNA | AA321649.1 | - | 2 | 209 | ||
| BC010954.1 | - | 2 | 174 | 4274617 | ||
| X02530.1 | - | 2 | 170 | |||