siRNA Details
Gene Symbol: C9orf129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098808.1 | NP_001092278.1 | 5 | 880 | ||
XM_006717117.3 | XP_006717180.1 | 4 | 899 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9orf129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098808.1 | NP_001092278.1 | 5 | 880 | ||
XM_006717117.3 | XP_006717180.1 | 4 | 899 |