siRNA Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305942.1 | NP_001292871.1 | 4 | 675 | ||
| NM_016035.4 | NP_057119.2 | 7 | 996 | |||
| GenBank mRNA | AF151850.1 | - | 7 | 967 | ||
| AK025376.1 | - | 7 | 910 | |||
| AK095083.1 | - | 3 | 2461 | |||
| AK128853.1 | - | 2 | 4660 | |||
| AK313650.1 | - | 7 | 979 | |||
| BC011895.2 | - | 7 | 666 | 3944239 | ||
| CB111694.1 | - | 4 | 346 | |||
| EU216419.1 | - | 7 | 978 | |||
| EU216420.1 | - | 7 | 896 | |||
| EU216421.1 | - | 7 | 730 | |||
| EU216422.1 | - | 7 | 668 | |||
| EU216423.1 | - | 6 | 592 | |||

COQ4
Genomic Map