siRNA Details
| Gene Symbol: ZNF100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173531.3 | NP_775802.2 | 5 | 1606 | ||
| GenBank mRNA | AK126551.1 | - | 5 | 1557 | ||
| BC031248.1 | - | 4 | 1571 | |||
| BC035579.1 | - | 5 | 1518 | 5576923 | ||
| BX640982.1 | - | 1 | 316 | |||

ZNF100
Genomic Map