siRNA Details
| Gene Symbol: C2orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105519.2 | NP_001098989.1 | 3 | 422 | ||
| NM_001322426.1 | NP_001309355.1 | 4 | 458 | |||
| XM_011532810.2 | XP_011531112.1 | 5 | 625 | |||
| XM_011532811.2 | XP_011531113.1 | 5 | 2121 | |||
| XM_017003962.1 | XP_016859451.1 | 5 | 476 | |||
| XM_017003963.1 | XP_016859452.1 | 3 | 420 | |||
| XM_017003964.1 | XP_016859453.1 | 4 | 452 | |||
| XM_017003965.1 | XP_016859454.1 | 5 | 476 | |||
| XM_017003966.1 | XP_016859455.1 | 5 | 625 | |||
| GenBank mRNA | AK091483.1 | - | 3 | 369 | ||
| BX114846.1 | - | 4 | 431 | |||
| HY021315.1 | - | 4 | 461 | |||

C2orf70
Genomic Map