siRNA Details
| Gene Symbol: C1orf210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164829.1 | NP_001158301.1 | 3 | 263 | ||
| NM_182517.2 | NP_872323.1 | 3 | 334 | |||
| XM_011540802.1 | XP_011539104.1 | 3 | 393 | |||
| GenBank mRNA | AL696898.1 | - | 3 | 264 | ||
| BC041633.1 | - | 3 | 323 | 4868093 | ||
| BM784450.1 | - | 3 | 334 | |||

C1orf210
Genomic Map