siRNA Details
| Gene Symbol: PRSS48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183375.2 | NP_899231.2 | 2 | 183 | ||
| XM_011531921.2 | XP_011530223.1 | 1 | 629 | |||
| GenBank mRNA | BC121109.1 | - | 2 | 187 | 40120369 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS48| Gene Symbol: PRSS48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183375.2 | NP_899231.2 | 2 | 183 | ||
| XM_011531921.2 | XP_011530223.1 | 1 | 629 | |||
| GenBank mRNA | BC121109.1 | - | 2 | 187 | 40120369 | |