siRNA Details
| Gene Symbol: ZSCAN30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112734.3 | NP_001106205.1 | 4 | 1547 | ||
| NM_001166012.2 | NP_001159484.1 | 5 | 1751 | |||
| NM_001288711.1 | NP_001275640.1 | 2 | 844 | |||
| XM_005258183.4 | XP_005258240.1 | 4 | 1516 | |||
| XM_006722371.3 | XP_006722434.1 | 5 | 1680 | |||
| XM_011525789.2 | XP_011524091.1 | 1 | 745 | |||
| XM_017025515.1 | XP_016881004.1 | 5 | 1699 | |||
| XM_017025516.1 | XP_016881005.1 | 4 | 1532 | |||
| XM_017025517.1 | XP_016881006.1 | 4 | 1526 | |||
| XM_017025518.1 | XP_016881007.1 | 4 | 1598 | |||
| XM_017025519.1 | XP_016881008.1 | 4 | 1503 | |||
| XM_017025520.1 | XP_016881009.1 | 3 | 1397 | |||
| XM_017025521.1 | XP_016881010.1 | 3 | 1350 | |||
| XM_017025522.1 | XP_016881011.1 | 3 | 1329 | |||
| GenBank mRNA | AF303374.1 | - | 1 | 969 | ||
| AK057386.1 | - | 1 | 300 | |||
| AK303447.1 | - | 5 | 1735 | |||
| AL832180.1 | - | 1 | 55 | |||
| AY234408.1 | - | 4 | 1510 | |||

ZSCAN30
Genomic Map