siRNA Details
| Gene Symbol: TMSB15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194324.2 | NP_919305.2 | 3 | 627 | ||
| GenBank mRNA | AK291479.1 | - | 3 | 603 | ||
| BC093093.1 | - | 3 | 627 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMSB15B| Gene Symbol: TMSB15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194324.2 | NP_919305.2 | 3 | 627 | ||
| GenBank mRNA | AK291479.1 | - | 3 | 603 | ||
| BC093093.1 | - | 3 | 627 | |||