siRNA Details
| Gene Symbol: TBC1D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015079.5 | NP_055894.6 | 6, 7 | 1471 | ||
| NM_144572.1 | NP_653173.1 | 6, 7 | 1471 | |||
| XM_017022022.1 | XP_016877511.1 | 8, 9 | 1492 | |||
| GenBank mRNA | AB028978.1 | - | 6, 7 | 1334 | ||
| BC033712.1 | - | 1, 2 | 254 | |||
| BC140766.1 | - | 5, 6 | 1116 | 9021634 | ||
| BC144482.1 | - | 5, 6 | 1116 | 9053009 | ||
| BC152465.1 | - | 6, 7 | 1334 | 8862601 | ||

TBC1D2B
Genomic Map