siRNA Details
| Gene Symbol: KCTD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100915.1 | NP_001094385.1 | 12 | 1984 | ||
| GenBank mRNA | AK302567.1 | - | 11 | 3202 | ||
| AK302748.1 | - | 12 | 1968 | |||
| BC070103.1 | - | 12 | 3765 | |||
| BC094790.1 | - | 10 | 3931 | |||
| BC117335.1 | - | 12 | 1984 | 40125886 | ||

KCTD19
Genomic Map