siRNA Details
| Gene Symbol: KDELC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153705.4 | NP_714916.3 | 2 | 286 | ||
| XM_011542621.2 | XP_011540923.1 | 2 | 259 | |||
| XM_011542623.2 | XP_011540925.1 | 2 | 253 | |||
| GenBank mRNA | AF533708.1 | - | 2 | 245 | ||
| AK125519.1 | - | 1 | 68 | |||
| AK313534.1 | - | 2 | 148 | |||
| AY358616.1 | - | 1 | 355 | |||
| BC036526.2 | - | 2 | 240 | 5266665 | ||
| DN999478.1 | - | 2 | 294 | |||

KDELC2
Genomic Map