siRNA Details
Gene Symbol: BLOC1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212550.4 | NP_997715.1 | 2 | 859 | ||
GenBank mRNA | AK130186.1 | - | 1 | 138 | ||
AK296695.1 | - | 2 | 857 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BLOC1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212550.4 | NP_997715.1 | 2 | 859 | ||
GenBank mRNA | AK130186.1 | - | 1 | 138 | ||
AK296695.1 | - | 2 | 857 |