siRNA Details
| Gene Symbol: BLOC1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212550.4 | NP_997715.1 | 2 | 676 | ||
| GenBank mRNA | AK296695.1 | - | 2 | 674 | ||
| AY531266.1 | - | 1 | 580 | |||
| BC151151.1 | - | 1 | 654 | 9021441 | ||
| BC151152.1 | - | 1 | 652 | 9021442 | ||

BLOC1S3
Genomic Map