siRNA Details
| Gene Symbol: FAM47E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136570.2 | NP_001130042.1 | 8 | 1183 | ||
| NM_001242936.1 | NP_001229865.1 | 8 | 1256 | |||
| GenBank mRNA | AK124936.1 | - | 8 | 1256 | ||
| BM713546.1 | - | 4 | 428 | |||
| CR627383.1 | - | 6 | 5859 | |||

FAM47E
Genomic Map