siRNA Details
| Gene Symbol: TNRC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024843.1 | NP_001020014.1 | 23 | 2880 | ||
| NM_001162501.1 | NP_001155973.1 | 22 | 5201 | |||
| NM_015088.2 | NP_055903.2 | 20 | 4871 | |||
| GenBank mRNA | AB029016.2 | - | 20 | 4851 | ||
| AK294519.1 | - | 22 | 5192 | |||
| AK304087.1 | - | 17 | 2356 | |||
| BC028626.2 | - | 23 | 2813 | 4826787 | ||
| BX647630.1 | - | 17 | 4073 | |||

TNRC6B
Genomic Map