siRNA Details
| Gene Symbol: TMEM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137560.1 | NP_001131032.1 | 2 | 304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM151B| Gene Symbol: TMEM151B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137560.1 | NP_001131032.1 | 2 | 304 | ||