siRNA Details
| Gene Symbol: INF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031714.3 | NP_001026884.3 | 4 | 687 | ||
| NM_022489.3 | NP_071934.3 | 4 | 687 | |||
| NM_032714.2 | NP_116103.1 | 4 | 687 | |||
| XM_005268004.4 | XP_005268061.1 | 4 | 774 | |||
| XM_005268005.4 | XP_005268062.1 | 4 | 745 | |||
| XM_017021595.1 | XP_016877084.1 | 4 | 745 | |||
| GenBank mRNA | AK290083.1 | - | 4 | 672 | ||
| BC006173.1 | - | 4 | 672 | |||
| BI489810.1 | - | 4 | 688 | |||
| BX248757.1 | - | 4 | 656 | |||

INF2
Genomic Map