siRNA Details
| Gene Symbol: KNCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001097611.1 | NP_001091080.1 | 3 | 695 | ||
| NM_001322255.1 | NP_001309184.1 | 4 | 1206 | |||
| GenBank mRNA | AK056573.1 | - | 3 | 623 | ||
| BC101295.1 | - | 3 | 617 | |||
| BC101296.1 | - | 3 | 617 | |||
| BC101297.2 | - | 3 | 617 | |||
| BC101298.2 | - | 3 | 617 | |||

KNCN
Genomic Map