siRNA Details
Gene Symbol: FCGR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201563.5 | NP_963857.3 | 7 | 1174 | ||
NR_047648.1 | 7 | 1174 | ||||
GenBank mRNA | X17652.1 | - | 7 | 1105 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FCGR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201563.5 | NP_963857.3 | 7 | 1174 | ||
NR_047648.1 | 7 | 1174 | ||||
GenBank mRNA | X17652.1 | - | 7 | 1105 |