siRNA Details
| Gene Symbol: PEX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000287.3 | NP_000278.3 | 2 | 1094 | ||
| NM_001316313.1 | NP_001303242.1 | 2 | 830 | |||
| NR_133009.1 | 2 | 1094 | ||||
| XM_011514661.1 | XP_011512963.1 | 2 | 1094 | |||
| GenBank mRNA | AB051076.1 | - | 2 | 1001 | ||
| AB051077.1 | - | 2 | 737 | |||
| AB051078.1 | - | 2 | 1001 | |||
| AB208906.1 | - | 2 | 1054 | |||
| AK225704.1 | - | 2 | 1032 | |||
| AK314237.1 | - | 2 | 1030 | |||
| BC033487.1 | - | 2 | 1009 | |||
| BC048331.1 | - | 2 | 1017 | 5189685 | ||

PEX6
Genomic Map