siRNA Details
Gene Symbol: TP73-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033710.1 | 4 | 2391 | |||
NR_033711.1 | 4 | 2391 | ||||
GenBank mRNA | AB007964.1 | - | 4 | 2341 | ||
AL133648.1 | - | 4 | 1644 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TP73-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033710.1 | 4 | 2391 | |||
NR_033711.1 | 4 | 2391 | ||||
GenBank mRNA | AB007964.1 | - | 4 | 2341 | ||
AL133648.1 | - | 4 | 1644 |