siRNA Details
| Gene Symbol: ZNF843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136509.1 | NP_001129981.1 | 2 | 1343 | ||
| GenBank mRNA | AK096248.1 | - | 2 | 1343 | ||
| AK291063.1 | - | 2 | 1101 | |||
| BC036762.1 | - | 2 | 969 | 5588928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF843| Gene Symbol: ZNF843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136509.1 | NP_001129981.1 | 2 | 1343 | ||
| GenBank mRNA | AK096248.1 | - | 2 | 1343 | ||
| AK291063.1 | - | 2 | 1101 | |||
| BC036762.1 | - | 2 | 969 | 5588928 | ||