siRNA Details
| Gene Symbol: ZNF717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128223.2 | NP_001121695.1 | 5 | 1163 | ||
| NM_001290208.1 | NP_001277137.1 | 5 | 1144 | |||
| NM_001290209.1 | NP_001277138.1 | 5 | 1173 | |||
| NM_001324027.1 | NP_001310956.1 | 5 | 1163 | |||
| XM_005264711.3 | XP_005264768.1 | 6 | 1241 | |||
| XM_006712912.3 | XP_006712975.1 | 5 | 1209 | |||
| XM_011533244.2 | XP_011531546.1 | 6 | 1091 | |||
| XM_011533245.2 | XP_011531547.1 | 6 | 1227 | |||
| XM_011533246.2 | XP_011531548.1 | 6 | 1092 | |||
| XM_011533248.2 | XP_011531550.1 | 6 | 1092 | |||
| XM_011533249.2 | XP_011531551.1 | 6 | 1090 | |||

ZNF717
Genomic Map