siRNA Details
Gene Symbol: CACNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000725.3 | NP_000716.2 | 7, 8 | 1032 | ||
NM_001206915.1 | NP_001193844.1 | 6, 7 | 909 | |||
NM_001206916.1 | NP_001193845.1 | 7, 8 | 683 | |||
NM_001206917.1 | NP_001193846.1 | 7, 8 | 650 | |||
XM_011538727.2 | XP_011537029.1 | 8, 9 | 768 | |||
XM_011538728.2 | XP_011537030.1 | 7, 8 | 1399 | |||
XM_017019957.1 | XP_016875446.1 | 8, 9 | 1325 | |||
XM_017019958.1 | XP_016875447.1 | 8, 9 | 1530 | |||
XM_017019959.1 | XP_016875448.1 | 5, 6 | 508 | |||
GenBank mRNA | AK122911.1 | - | 6, 7 | 2105 | ||
AK289709.1 | - | 7, 8 | 699 | |||
AK297639.1 | - | 7, 8 | 670 | |||
AK304537.1 | - | 7, 8 | 631 | |||
AK316323.1 | - | 7, 8 | 666 | |||
BC041811.1 | - | 7, 8 | 772 | 5315091 | ||
CD013906.1 | - | 6, 7 | 471 | |||
L27584.1 | - | 7, 8 | 597 | |||
U07139.1 | - | 7, 8 | 607 | |||
X76555.1 | - | 7, 8 | 611 | |||
X76556.1 | - | 7, 8 | 600 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
