siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 13 | 1263 | ||
| XM_005260684.3 | XP_005260741.1 | 13 | 1257 | |||
| XM_017027783.1 | XP_016883272.1 | 10 | 884 | |||
| GenBank mRNA | AK226100.1 | - | 8 | 687 | ||
| AK303375.1 | - | 4 | 632 | |||
| AK310540.1 | - | 13 | 1281 | |||
| AL110249.1 | - | 4 | 228 | |||
| BC122529.1 | - | 4 | 481 | 40012183 | ||

C20orf194
Genomic Map