siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 4, 5 | 505 | ||
| XM_005260684.3 | XP_005260741.1 | 4, 5 | 502 | |||
| XM_017027783.1 | XP_016883272.1 | 2, 3 | 182 | |||
| GenBank mRNA | AK310540.1 | - | 4, 5 | 523 | ||
| AU311821.1 | - | 4, 5 | 441 | |||
| CN285135.1 | - | 2, 3 | 152 | |||

C20orf194
Genomic Map