siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 7 | 731 | ||
| XM_005260684.3 | XP_005260741.1 | 7 | 728 | |||
| XM_017027783.1 | XP_016883272.1 | 5 | 408 | |||
| GenBank mRNA | AK226100.1 | - | 3 | 211 | ||
| AK310540.1 | - | 7 | 749 | |||
| BX424937.2 | - | 7 | 677 | |||
| CN285135.1 | - | 5 | 378 | |||

C20orf194
Genomic Map