siRNA Details
| Gene Symbol: CFAP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304548.1 | NP_001291477.1 | 62 | 9166 | ||
| GenBank mRNA | BG197183.1 | - | 2 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP47| Gene Symbol: CFAP47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304548.1 | NP_001291477.1 | 62 | 9166 | ||
| GenBank mRNA | BG197183.1 | - | 2 | 254 | ||