siRNA Details
| Gene Symbol: DRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171184.1 | NP_001164655.1 | 6 | 957 | ||
| NM_001939.2 | NP_001930.2 | 8 | 1240 | |||
| XM_017029333.1 | XP_016884822.1 | 8 | 1398 | |||
| XM_017029334.1 | XP_016884823.1 | 8 | 1398 | |||
| GenBank mRNA | AK289825.1 | - | 8 | 1128 | ||
| AK295843.1 | - | 6 | 826 | |||
| BC111695.1 | - | 8 | 1077 | 40036070 | ||
| U43519.1 | - | 8 | 1189 | |||

DRP2
Genomic Map