siRNA Details
| Gene Symbol: DRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171184.1 | NP_001164655.1 | 8 | 1185 | ||
| NM_001939.2 | NP_001930.2 | 10 | 1468 | |||
| XM_017029333.1 | XP_016884822.1 | 10 | 1626 | |||
| XM_017029334.1 | XP_016884823.1 | 9 | 1547 | |||
| GenBank mRNA | AK289825.1 | - | 10 | 1356 | ||
| AK295843.1 | - | 8 | 1054 | |||
| BC111695.1 | - | 10 | 1305 | 40036070 | ||
| U43519.1 | - | 10 | 1417 | |||

DRP2
Genomic Map