siRNA Details
| Gene Symbol: PLXND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015103.2 | NP_055918.2 | 19, 20 | 3849 | ||
| XM_011512588.1 | XP_011510890.1 | 19, 20 | 3764 | |||
| XM_011512589.1 | XP_011510891.1 | 16, 17 | 3374 | |||
| XM_011512590.2 | XP_011510892.1 | 19, 20 | 3764 | |||
| XM_011512592.1 | XP_011510894.1 | 7, 8 | 932 | |||
| GenBank mRNA | AB014520.2 | - | 19, 20 | 3928 | ||
| AY116661.1 | - | 19, 20 | 3929 | |||
| BC150280.1 | - | 19, 20 | 3928 | 9007263 | ||
| BU527983.1 | - | 1, 2 | 106 | |||

PLXND1
Genomic Map