siRNA Details
| Gene Symbol: GPATCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002909.3 | NP_001002909.1 | 2, 3 | 349 | ||
| XM_011524557.1 | XP_011522859.1 | 2, 3 | 336 | |||
| XM_017024375.1 | XP_016879864.1 | 2, 3 | 333 | |||
| GenBank mRNA | AK025600.1 | - | 2, 3 | 151 | ||
| BC140010.1 | - | 2, 3 | 151 | |||
| CD709791.1 | - | 2, 3 | 213 | |||
| DB021837.1 | - | 2, 3 | 336 | |||
| HY048299.1 | - | 2, 3 | 350 | |||

GPATCH8
Genomic Map