siRNA Details
| Gene Symbol: KDM6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080424.1 | NP_001073893.1 | 12 | 3872 | ||
| XM_005256549.3 | XP_005256606.1 | 13 | 4024 | |||
| XM_005256550.4 | XP_005256607.1 | 12 | 3866 | |||
| XM_005256551.3 | XP_005256608.1 | 12 | 3905 | |||
| XM_005256552.4 | XP_005256609.1 | 12 | 4878 | |||
| XM_006721483.3 | XP_006721546.1 | 13 | 3969 | |||
| XM_011523750.2 | XP_011522052.1 | 13 | 5229 | |||
| XM_011523752.2 | XP_011522054.1 | 12 | 3869 | |||
| XM_017024381.1 | XP_016879870.1 | 12 | 5315 | |||
| GenBank mRNA | AB002344.2 | - | 12 | 3877 | ||
| BC009994.1 | - | 2 | 501 | |||

KDM6B
Genomic Map