siRNA Details
| Gene Symbol: ZZEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015113.3 | NP_055928.3 | 42 | 6849 | ||
| XM_005256561.2 | XP_005256618.1 | 42 | 6913 | |||
| XM_017024382.1 | XP_016879871.1 | 42 | 6913 | |||
| XM_017024383.1 | XP_016879872.1 | 42 | 6910 | |||
| XM_017024384.1 | XP_016879873.1 | 17 | 2724 | |||
| GenBank mRNA | AB007859.4 | - | 42 | 6849 | ||
| AK001683.1 | - | 13 | 1948 | |||
| BC047285.1 | - | 13 | 2111 | |||
| BC151836.1 | - | 42 | 6849 | 8860131 | ||

ZZEF1
Genomic Map