siRNA Details
| Gene Symbol: ZZEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015113.3 | NP_055928.3 | 35 | 5604 | ||
| XM_005256561.2 | XP_005256618.1 | 35 | 5668 | |||
| XM_017024382.1 | XP_016879871.1 | 35 | 5668 | |||
| XM_017024383.1 | XP_016879872.1 | 35 | 5665 | |||
| XM_017024384.1 | XP_016879873.1 | 10 | 1479 | |||
| GenBank mRNA | AB007859.4 | - | 35 | 5604 | ||
| AK001683.1 | - | 6 | 703 | |||
| AK122719.1 | - | 10 | 1698 | |||
| BC047285.1 | - | 6 | 866 | |||
| BC151836.1 | - | 35 | 5604 | 8860131 | ||

ZZEF1
Genomic Map