siRNA Details
| Gene Symbol: SSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198455.2 | NP_940857.2 | 57 | 8670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSPO| Gene Symbol: SSPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198455.2 | NP_940857.2 | 57 | 8670 | ||