siRNA Details
| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019814.4 | NP_062788.1 | 3 | 649 | ||
| GenBank mRNA | AB253941.1 | - | 2 | 161 | ||
| AF141312.1 | - | 3 | 180 | |||
| AK004166.1 | - | 3 | 276 | |||
| AK008756.1 | - | 3 | 276 | |||
| AK018360.1 | - | 3 | 277 | |||
| AY028386.1 | - | 3 | 202 | |||
| AY028387.1 | - | 3 | 187 | |||
| BC021594.1 | - | 3 | 199 | 5148784 | ||
| BC090259.1 | - | 3 | 253 | 5060413 | ||
| BC132371.1 | - | 3 | 267 | 40130648 | ||
| BC151191.1 | - | 3 | 267 | 8861324 | ||
| BE457620.1 | - | 3 | 445 | |||

Higd1a
Genomic Map