siRNA Details
| Gene Symbol: Lrrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172879.3 | NP_766467.2 | 2 | 1041 | ||
| GenBank mRNA | AK029981.1 | - | 2 | 1042 | ||
| BC141026.1 | - | 2 | 868 | 9055638 | ||
| BC145170.1 | - | 2 | 868 | 9053721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrd1| Gene Symbol: Lrrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172879.3 | NP_766467.2 | 2 | 1041 | ||
| GenBank mRNA | AK029981.1 | - | 2 | 1042 | ||
| BC141026.1 | - | 2 | 868 | 9055638 | ||
| BC145170.1 | - | 2 | 868 | 9053721 | ||