siRNA Details
| Gene Symbol: 1700003F12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029305.2 | NP_083581.1 | 3 | 678 | ||
| GenBank mRNA | BC039797.1 | - | 3 | 540 | ||
| BC147532.1 | - | 3 | 666 | 9056523 | ||
| BC147533.1 | - | 3 | 666 | 9056524 | ||
| DQ080431.1 | - | 3 | 679 | |||

1700003F12Rik
Genomic Map