siRNA Details
| Gene Symbol: Cnfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081375.1 | NP_001074844.1 | 4 | 375 | ||
| NM_028219.1 | NP_082495.1 | 5 | 568 | |||
| GenBank mRNA | AK008979.1 | - | 4 | 376 | ||
| BC059093.1 | - | 5 | 547 | 30293557 | ||

Cnfn
Genomic Map