siRNA Details
| Gene Symbol: Dock10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285927.1 | NP_001272856.1 | 26 | 3030 | ||
| NM_175291.4 | NP_780500.2 | 26 | 3014 | |||
| GenBank mRNA | AK122347.1 | - | 17 | 1868 | ||
| BC150753.1 | - | 26 | 2950 | 9087664 | ||
| GU797481.1 | - | 26 | 2987 | |||

Dock10
Genomic Map