siRNA Details
| Gene Symbol: C330007P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029951.1 | NP_084227.1 | 4 | 507 | ||
| GenBank mRNA | AK021190.1 | - | 4 | 500 | ||
| AK032304.1 | - | 4 | 422 | |||
| AK034943.1 | - | 4 | 448 | |||
| AK035825.1 | - | 4 | 496 | |||
| AK036910.1 | - | 4 | 430 | |||
| AK044209.1 | - | 4 | 444 | |||
| AK086009.1 | - | 1 | 2701 | |||
| AK141615.1 | - | 4 | 448 | |||
| AK184450.1 | - | 1 | 98 | |||
| BB622061.1 | - | 4 | 508 | |||
| BC021479.1 | - | 652 | 5344198 | |||
| BC051144.1 | - | 4 | 420 | 6588980 | ||
| BC085302.1 | - | 4 | 395 | |||

C330007P06Rik
Genomic Map