siRNA Details
| Gene Symbol: Gm13242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103158.2 | NP_001096628.2 | 5 | 1387 | ||
| GenBank mRNA | BC096593.1 | - | 5 | 1399 | 30459933 | |
| Gene Symbol: Zfp600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 1966 | ||
| NM_001177546.2 | NP_001171017.1 | 5 | 2059 | |||
| GenBank mRNA | BC108352.1 | - | 5 | 1966 | 30462028 | |

Gm13242
Genomic Map