siRNA Details
| Gene Symbol: Rell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153793.2 | NP_722488.2 | 5, 6 | 910 | ||
| GenBank mRNA | AK044097.1 | - | 5, 6 | 873 | ||
| BC038500.1 | - | 5, 6 | 853 | 4481989 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rell2| Gene Symbol: Rell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153793.2 | NP_722488.2 | 5, 6 | 910 | ||
| GenBank mRNA | AK044097.1 | - | 5, 6 | 873 | ||
| BC038500.1 | - | 5, 6 | 853 | 4481989 | ||