siRNA Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033606.2 | NP_001028778.2 | 2 | 647 | ||
| NM_028817.3 | NP_083093.2 | 3 | 787 | |||
| GenBank mRNA | AK012088.1 | - | 2 | 590 | ||
| AK161708.1 | - | 3 | 788 | |||
| AK166723.1 | - | 3 | 726 | |||

Acsl3
Genomic Map