siRNA Details
| Gene Symbol: Map4k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290345.1 | NP_001277274.1 | 3 | 360 | ||
| GenBank mRNA | AI430883.1 | - | 3 | 360 | ||
| AK014711.1 | - | 3 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map4k3| Gene Symbol: Map4k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290345.1 | NP_001277274.1 | 3 | 360 | ||
| GenBank mRNA | AI430883.1 | - | 3 | 360 | ||
| AK014711.1 | - | 3 | 349 | |||