siRNA Details
| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024413.2 | NP_077724.2 | 2 | 272 | ||
| GenBank mRNA | AK143976.1 | - | 261 | |||
| AK157990.1 | - | 2 | 261 | |||
| AK170283.1 | - | 2 | 271 | |||
| AK171398.1 | - | 2 | 261 | |||
| AW227287.1 | - | 2 | 272 | |||

Plekhf1
Genomic Map