siRNA Details
| Gene Symbol: Plekhf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024413.2 | NP_077724.2 | 2 | 1518 | ||
| GenBank mRNA | AK007489.1 | - | 1 | 926 | ||
| AK143976.1 | - | 1507 | ||||
| AK157990.1 | - | 2 | 1507 | |||
| AK170283.1 | - | 2 | 1517 | |||
| BC002120.1 | - | 1 | 1484 | 3491671 | ||

Plekhf1
Genomic Map